Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258839.2 | NP_001245768.1 | 3 | 5798 | 146 | |
NM_001298537.1 | NP_001285466.1 | 1 | 7036 | 146 | ||
NM_134512.5 | NP_608356.1 | 1 | 7441 | 146 | ||
NM_167685.3 | NP_728304.1 | 2 | 7096 | 146 | ||
GenBank mRNA | BT132946.1 | - | 1 | 6489 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |