Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dop2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014757.3 | NP_001014757.2 | 5 - 6 | 2251 | 96 | |
NM_001014758.3 | NP_001014758.2 | 5 - 6 | 2251 | 96 | ||
NM_001014759.2 | NP_001014759.1 | 3 - 4 | 738 | 96 | ||
NM_001014760.3 | NP_001014760.2 | 5 - 6 | 2251 | 96 | ||
NM_001031909.2 | NP_001027080.2 | 5 - 6 | 2251 | 96 | ||
NM_001298548.1 | NP_001285477.1 | 4 - 5 | 2135 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |