Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obst-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258849.2 | NP_001245778.1 | 3 - 4 | 577 | 92 | |
NM_134534.3 | NP_608378.2 | 3 - 4 | 577 | 92 | ||
GenBank mRNA | BT001512.1 | - | 3 - 4 | 536 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |