Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RunxA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042820.3 | NP_001036285.2 | 2 - 3 | 1508 | 74 | |
NM_001272818.1 | NP_001259747.1 | 2 - 3 | 1508 | 74 | ||
NM_134556.2 | NP_608400.2 | 2 - 3 | 1508 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |