Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298570.1 | NP_001285499.1 | 2 - 3 | 1535 | 64 | |
NM_001298571.1 | NP_001285500.1 | 1 - 2 | 1296 | 64 | ||
NM_134559.3 | NP_608403.1 | 1 - 2 | 1761 | 64 | ||
GenBank mRNA | BT031316.1 | - | 1 - 2 | 933 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |