Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 14 - 15 | 5719 | 67 | |
NM_001103575.2 | NP_001097045.1 | 12 - 13 | 3061 | 67 | ||
NM_001258876.2 | NP_001245805.1 | 10 - 11 | 4864 | 67 | ||
NM_001258877.2 | NP_001245806.1 | 13 - 14 | 5297 | 67 | ||
NM_001258878.2 | NP_001245807.1 | 11 - 12 | 2631 | 67 | ||
NM_001258879.2 | NP_001245808.1 | 13 - 14 | 5980 | 67 | ||
NM_164353.2 | NP_722589.1 | 12 - 13 | 3064 | 67 | ||
NM_164354.3 | NP_722590.2 | 13 - 14 | 5941 | 67 | ||
GenBank mRNA | BT057995.1 | - | 12 - 13 | 3039 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |