Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 10 - 11 | 4665 | 95 | |
NM_001103575.2 | NP_001097045.1 | 8 - 9 | 2004 | 95 | ||
NM_001258876.2 | NP_001245805.1 | 6 - 7 | 3810 | 95 | ||
NM_001258877.2 | NP_001245806.1 | 9 - 10 | 4243 | 95 | ||
NM_001258878.2 | NP_001245807.1 | 7 - 8 | 1577 | 95 | ||
NM_001258879.2 | NP_001245808.1 | 9 - 10 | 4926 | 95 | ||
NM_164353.2 | NP_722589.1 | 8 - 9 | 2010 | 95 | ||
NM_164354.3 | NP_722590.2 | 9 - 10 | 4887 | 95 | ||
GenBank mRNA | BT057995.1 | - | 8 - 9 | 1985 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |