Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosbeta4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273021.1 | NP_001259950.1 | 1 - 2 | 158 | 77 | |
NM_164491.3 | NP_722823.2 | 1 - 2 | 158 | 77 | ||
GenBank mRNA | AY119488.1 | - | 1 - 2 | 158 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |