Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obst-E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164659.3 | NP_723116.1 | 1 - 2 | 181 | 89 | |
GenBank mRNA | BT099959.1 | - | 1 - 2 | 161 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |