Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyc32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103678.2 | NP_001097148.1 | 14 - 15 | 1958 | 89 | |
NM_001298905.1 | NP_001285834.1 | 13 - 14 | 1850 | 89 | ||
NM_176013.2 | NP_788027.1 | 13 - 14 | 1874 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |