Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIIb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144360.2 | NP_001137832.1 | 3 - 4 | 1381 | 80 | |
NM_176057.3 | NP_788071.3 | 3 - 4 | 1381 | 80 | ||
GenBank mRNA | BT025892.1 | - | 3 - 4 | 1411 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |