Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIIb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144360.2 | NP_001137832.1 | 6 - 7 | 1920 | 59 | |
NM_176057.3 | NP_788071.3 | 6 - 7 | 1920 | 59 | ||
GenBank mRNA | BT025892.1 | - | 6 - 7 | 1950 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |