Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043067.2 | NP_001036532.1 | 5 - 6 | 1567 | 80 | |
NM_001299284.1 | NP_001286213.1 | 5 - 6 | 1567 | 80 | ||
NM_176106.2 | NP_788286.1 | 5 - 6 | 1567 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |