Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103786.2 | NP_001097256.1 | 5 - 6 | 1014 | 71 | |
NM_001259307.3 | NP_001246236.1 | 6 - 7 | 2846 | 71 | ||
NM_001273925.2 | NP_001260854.1 | 6 - 7 | 2852 | 71 | ||
NM_001347757.1 | NP_001334718.1 | 5 - 6 | 3247 | 71 | ||
NM_176121.3 | NP_788301.1 | 5 - 6 | 3247 | 71 | ||
NM_176123.5 | NP_788303.1 | 6 - 7 | 2852 | 71 | ||
GenBank mRNA | BT133118.1 | - | 5 - 6 | 844 | 71 | |
M90806.1 | - | 5 - 6 | 71 |
Gene Symbol: CG12912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299344.1 | NP_001286273.1 | 5 - 6 | 3247 | 71 |
Gene Symbol: CG46321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347803.1 | NP_001334719.1 | 6 - 7 | 2852 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |