Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259307.3 | NP_001246236.1 | 2 - 3 | 2179 | 70 | |
NM_001273925.2 | NP_001260854.1 | 2 - 3 | 2185 | 70 | ||
NM_176123.5 | NP_788303.1 | 2 - 3 | 2185 | 70 | ||
GenBank mRNA | BT133118.1 | - | 1 - 2 | 177 | 70 | |
M90806.1 | - | 1 - 2 | 358 | 70 |
Gene Symbol: CG46321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347803.1 | NP_001334719.1 | 2 - 3 | 2185 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |