Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176272.2 | NP_788450.1 | 4 - 5 | 480 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |