Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169885.2 | NP_001163356.1 | 14 - 15 | 2162 | 66 | |
NM_001169886.2 | NP_001163357.1 | 9 - 10 | 2396 | 66 | ||
NM_001169887.2 | NP_001163358.1 | 15 - 16 | 2197 | 66 | ||
NM_001169888.2 | NP_001163359.1 | 11 - 12 | 2501 | 66 | ||
NM_001169890.2 | NP_001163361.1 | 9 - 10 | 1359 | 66 | ||
NM_001169891.2 | NP_001163362.2 | 14 - 15 | 2142 | 66 | ||
NM_001169892.2 | NP_001163363.1 | 13 - 14 | 2056 | 66 | ||
NM_001169893.2 | NP_001163364.1 | 14 - 15 | 2167 | 66 | ||
NM_001259722.1 | NP_001246651.1 | 14 - 15 | 2137 | 66 | ||
NM_001300031.1 | NP_001286960.1 | 9 - 10 | 2396 | 66 | ||
NM_001300032.1 | NP_001286961.1 | 14 - 15 | 2167 | 66 | ||
NM_001300033.1 | NP_001286962.1 | 12 - 13 | 2062 | 66 | ||
NM_176293.3 | NP_788471.1 | 9 - 10 | 2396 | 66 | ||
NM_176294.5 | NP_788472.2 | 9 - 10 | 2396 | 66 | ||
GenBank mRNA | AY118310.1 | - | 9 - 10 | 2371 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |