Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176295.2 | NP_788473.1 | 3 - 4 | 904 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |