Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176357.2 | NP_788535.2 | 4 - 5 | 1336 | 75 | |
NM_176358.2 | NP_788536.1 | 4 - 5 | 1339 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |