Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176365.3 | NP_788543.1 | 5 - 6 | 617 | 60 | |
GenBank mRNA | AY071612.1 | - | 5 - 6 | 581 | 60 | |
U15735.1 | - | 3 - 4 | 373 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |