Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdgC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370029.1 | NP_001356983.1 | 2 - 3 | 455 | 65 | |
NM_080490.4 | NP_536738.3 | 3 - 4 | 1304 | 65 | ||
NM_176366.2 | NP_788544.2 | 3 - 4 | 942 | 65 | ||
NM_176367.3 | NP_788545.3 | 3 - 4 | 1117 | 65 | ||
NM_176368.2 | NP_788546.1 | 2 - 3 | 632 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |