Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201665.2 | NP_001188594.1 | 14 - 15 | 3240 | 74 | |
NM_001201666.2 | NP_001188595.1 | 14 - 15 | 3240 | 74 | ||
NM_001201667.2 | NP_001188596.1 | 14 - 15 | 3240 | 74 | ||
NM_001201668.2 | NP_001188597.1 | 13 - 14 | 3302 | 74 | ||
NM_001298379.1 | NP_001285308.1 | 14 - 15 | 3432 | 74 | ||
NM_176740.4 | NP_788913.2 | 14 - 15 | 3240 | 74 | ||
GenBank mRNA | AF222744.1 | - | 14 - 15 | 3222 | 74 | |
BT021400.1 | - | 14 - 15 | 3215 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |