Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057228.5 | NP_476576.1 | 10 - 11 | 2296 | 67 | |
NM_057230.6 | NP_476578.3 | 10 - 11 | 2251 | 67 | ||
GenBank mRNA | AF032865.1 | - | 10 - 11 | 1390 | 67 | |
AF035825.1 | - | 10 - 11 | 2272 | 67 | ||
AF036584.1 | - | 9 - 10 | 1432 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |