Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300640.1 | NP_001287569.1 | 5 - 6 | 539 | 62 | |
NM_057292.4 | NP_476640.1 | 5 - 6 | 539 | 62 | ||
GenBank mRNA | K01567.1 | - | 5 - 6 | 517 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |