Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300663.1 | NP_001287592.1 | 1 - 2 | 724 | 95 | |
NM_057303.5 | NP_476651.1 | 1 - 2 | 344 | 95 | ||
GenBank mRNA | AY058596.1 | - | 1 - 2 | 322 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |