Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169248.4 | NP_731295.1 | 2 - 3 | 972 | 70 | |
NM_206464.3 | NP_996186.1 | 2 - 3 | 972 | 70 | ||
GenBank mRNA | AY058560.1 | - | 2 - 3 | 948 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |