Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276161.1 | NP_001263090.1 | 13 - 14 | 3117 | 74 | |
NM_057420.4 | NP_476768.1 | 13 - 14 | 3097 | 74 | ||
GenBank mRNA | BT150384.1 | - | 13 - 14 | 3099 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |