Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276161.1 | NP_001263090.1 | 6 - 7 | 1393 | 84 | |
NM_057420.4 | NP_476768.1 | 6 - 7 | 1373 | 84 | ||
GenBank mRNA | BT150384.1 | - | 6 - 7 | 1375 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |