Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276214.1 | NP_001263143.1 | 11 - 12 | 3144 | 69 | |
NM_057519.4 | NP_476867.1 | 12 - 13 | 3192 | 69 | ||
GenBank mRNA | BT031258.1 | - | 12 - 13 | 2656 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |