Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276214.1 | NP_001263143.1 | 6 - 7 | 2067 | 115 | |
NM_057519.4 | NP_476867.1 | 7 - 8 | 2115 | 115 | ||
GenBank mRNA | BT031258.1 | - | 7 - 8 | 1579 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |