Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm02134819_g1
See other TMOD GE Assays ›
Gene Symbol:
tmod
Gene Name:
tropomodulin
Gene Aliases:
1309/10, CG11493, CG1539, CG15540, CT3919, CT41421, Dmel\CG1539, Dmel_CG1539, E42, Tmod, l(3)00848, l(3)S130910, spdo
Chromosome Location:
Chr.3R: 30532607 - 30578905 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0082582
Assay ID
Dm02134819_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: tmod
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001104512.4
NP_001097982.1
5 - 6
1371
67
NM_001170304.3
NP_001163775.1
4 - 5
1403
67
NM_001170305.3
NP_001163776.1
5 - 6
1686
67
NM_001170306.2
NP_001163777.1
5 - 6
1439
67
NM_001202390.1
NP_001189319.1
5 - 6
1401
67
NM_001202391.2
NP_001189320.1
5 - 6
1348
67
NM_001260442.2
NP_001247371.1
5 - 6
1365
67
NM_001260443.2
NP_001247372.1
5 - 6
1479
67
NM_001260444.2
NP_001247373.1
5 - 6
1432
67
NM_001276182.1
NP_001263111.1
4 - 5
1351
67
NM_001276183.1
NP_001263112.1
4 - 5
1351
67
NM_001382033.1
NP_001369033.1
5 - 6
1371
67
NM_057683.5
NP_477031.1
4 - 5
1223
67
NM_057684.4
NP_477032.1
4 - 5
1194
67
NM_170503.4
NP_733382.1
4 - 5
1592
67
NM_170504.4
NP_733383.1
5 - 6
1371
67
NM_170505.4
NP_733384.1
5 - 6
1456
67
NM_170506.3
NP_733385.1
5 - 6
1402
67
GenBank mRNA
AY094727.1
-
4 - 5
1194
67
BT099728.1
-
5 - 6
1402
67
BT100046.1
-
5 - 6
1440
67
BT133205.1
-
4 - 5
1195
67
BT133473.1
-
5 - 6
1364
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Drosophila melanogaster
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
pointed-end actin filament capping
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0