Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057874.5 | NP_477222.1 | 1 - 2 | 265 | 93 | |
GenBank mRNA | AY118406.1 | - | 1 - 2 | 272 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |