Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014614.2 | NP_001014614.1 | 3 - 4 | 438 | 116 | |
NM_001260099.1 | NP_001247028.1 | 3 - 4 | 396 | 116 | ||
NM_057906.4 | NP_477254.1 | 2 - 3 | 376 | 116 | ||
NM_169350.3 | NP_731489.2 | 3 - 4 | 551 | 116 | ||
NM_169351.2 | NP_731490.1 | 3 - 4 | 322 | 116 | ||
GenBank mRNA | BT125950.1 | - | 3 - 4 | 421 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |