Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057923.4 | NP_477271.1 | 1 - 2 | 1966 | 82 | |
GenBank mRNA | AY129448.1 | - | 1 - 2 | 1981 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |