Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057970.4 | NP_477318.1 | 7 - 8 | 1403 | 76 | |
GenBank mRNA | AY119509.1 | - | 7 - 8 | 1361 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |