Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058133.5 | NP_477481.1 | 1 - 2 | 507 | 65 | |
GenBank mRNA | AF260235.1 | - | 1 - 2 | 432 | 65 | |
AY051703.1 | - | 1 - 2 | 432 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |