Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259980.2 | NP_001246909.1 | 1 - 2 | 348 | 93 | |
NM_079501.4 | NP_524225.1 | 1 - 2 | 348 | 93 | ||
GenBank mRNA | AF043264.1 | - | 1 - 2 | 265 | 93 | |
Y16240.1 | - | 1 - 2 | 283 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |