Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104217.2 | NP_001097687.1 | 1 - 2 | 315 | 77 | |
NM_079511.5 | NP_524235.2 | 2 - 3 | 501 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |