Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104217.2 | NP_001097687.1 | 3 - 4 | 1063 | 62 | |
NM_079511.5 | NP_524235.2 | 4 - 5 | 1249 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |