Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300253.1 | NP_001287182.1 | 3 - 4 | 820 | 73 | |
NM_079513.3 | NP_524237.1 | 4 - 5 | 722 | 73 | ||
NM_169073.2 | NP_730957.1 | 3 - 4 | 820 | 73 | ||
GenBank mRNA | AF145618.1 | - | 3 - 4 | 765 | 73 | |
U38198.1 | - | 3 - 4 | 654 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |