Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104227.3 | NP_001097697.2 | 4 - 5 | 2909 | 69 | |
NM_001104228.3 | NP_001097698.1 | 4 - 5 | 3932 | 69 | ||
NM_001170065.2 | NP_001163536.1 | 4 - 5 | 2909 | 69 | ||
NM_001170066.2 | NP_001163537.1 | 4 - 5 | 2909 | 69 | ||
NM_001275399.1 | NP_001262328.1 | 8 - 9 | 4059 | 69 | ||
NM_001275400.1 | NP_001262329.1 | 8 - 9 | 3876 | 69 | ||
NM_001275401.1 | NP_001262330.1 | 4 - 5 | 2909 | 69 | ||
NM_001275402.1 | NP_001262331.1 | 4 - 5 | 2909 | 69 | ||
NM_001275403.1 | NP_001262332.1 | 8 - 9 | 5055 | 69 | ||
NM_079526.3 | NP_524250.3 | 8 - 9 | 4032 | 69 | ||
NM_169170.3 | NP_731131.2 | 4 - 5 | 2909 | 69 | ||
NM_169171.2 | NP_731132.1 | 8 - 9 | 3849 | 69 | ||
NM_169172.3 | NP_731133.1 | 4 - 5 | 2909 | 69 | ||
GenBank mRNA | AF217960.1 | - | 8 - 9 | 4005 | 69 | |
AY051905.1 | - | 4 - 5 | 2872 | 69 | ||
BT149892.1 | - | 8 - 9 | 3996 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |