Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300306.1 | NP_001287235.1 | 1 - 2 | 159 | 62 | |
NM_079558.5 | NP_524282.4 | 1 - 2 | 159 | 62 | ||
GenBank mRNA | AY094800.1 | - | 1 - 2 | 102 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |