Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sodh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079587.3 | NP_524311.1 | 3 - 4 | 631 | 59 | |
GenBank mRNA | AY058731.1 | - | 3 - 4 | 619 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |