Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079620.3 | NP_524344.1 | 2 - 3 | 975 | 65 | |
GenBank mRNA | AY071489.1 | - | 2 - 3 | 973 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |