Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or88a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079624.3 | NP_524348.2 | 2 - 3 | 1002 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |