Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170148.2 | NP_001163619.1 | 5 - 6 | 583 | 87 | |
NM_001260191.1 | NP_001247120.1 | 3 - 4 | 833 | 87 | ||
NM_001382175.1 | NP_001369014.1 | 5 - 6 | 870 | 87 | ||
NM_079636.4 | NP_524360.2 | 3 - 4 | 835 | 87 | ||
NM_206494.3 | NP_996216.1 | 5 - 6 | 583 | 87 | ||
GenBank mRNA | AY060414.1 | - | 3 - 4 | 815 | 87 | |
BT011469.1 | - | 5 - 6 | 581 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |