Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079642.3 | NP_524366.1 | 2 - 3 | 1246 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |