Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cher | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275728.1 | NP_001262657.2 | 2 - 3 | 597 | 57 | |
NM_001275730.1 | NP_001262659.2 | 2 - 3 | 597 | 57 | ||
NM_079659.4 | NP_524383.3 | 1 - 2 | 348 | 57 | ||
NM_206502.3 | NP_996224.2 | 1 - 2 | 348 | 57 | ||
GenBank mRNA | AF188360.1 | - | 1 - 2 | 324 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |