Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079674.3 | NP_524398.1 | 1 - 2 | 161 | 85 | |
GenBank mRNA | BT150161.1 | - | 1 - 2 | 162 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |