Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043257.2 | NP_001036722.1 | 4 - 5 | 4073 | 57 | |
NM_001043258.2 | NP_001036723.1 | 4 - 5 | 4073 | 57 | ||
NM_001043259.2 | NP_001036724.1 | 4 - 5 | 4115 | 57 | ||
NM_001043261.2 | NP_001036726.1 | 3 - 4 | 3786 | 57 | ||
NM_001043263.2 | NP_001036728.1 | 3 - 4 | 4000 | 57 | ||
NM_001275801.1 | NP_001262730.1 | 3 - 4 | 4000 | 57 | ||
NM_001275803.1 | NP_001262732.1 | 4 - 5 | 4115 | 57 | ||
GenBank mRNA | AF277386.1 | - | 3 - 4 | 3684 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |